missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Tolyldiethanolamine 98.0+%, TCI America™
$63.38 - $370.95
Chemical Identifiers
| CAS | 3077-12-1 |
|---|---|
| Molecular Formula | C11H17NO2 |
| Molecular Weight (g/mol) | 195.26 |
| MDL Number | MFCD00020580 |
| InChI Key | JUVSRZCUMWZBFK-UHFFFAOYSA-N |
| PubChem CID | 76497 |
| IUPAC Name | 2-[N-(2-hydroxyethyl)-4-methylanilino]ethanol |
| SMILES | CC1=CC=C(C=C1)N(CCO)CCO |
Chemical Identifiers
| 3077-12-1 | |
| 195.26 | |
| JUVSRZCUMWZBFK-UHFFFAOYSA-N | |
| 2-[N-(2-hydroxyethyl)-4-methylanilino]ethanol |
| C11H17NO2 | |
| MFCD00020580 | |
| 76497 | |
| CC1=CC=C(C=C1)N(CCO)CCO |
Specifications
| 3077-12-1 | |
| White-Yellow | |
| C11H17NO2 | |
| 25 g | |
| JUVSRZCUMWZBFK-UHFFFAOYSA-N | |
| 2-[N-(2-hydroxyethyl)-4-methylanilino]ethanol | |
| 76497 | |
| ≥98.0% (GC) | |
| p-Tolyldiethanolamine |
| 52°C | |
| 163°C | |
| MFCD00020580 | |
| N,N-Bis(2-hydroxyethyl)-p-toluidine, 2,2′C-(4-Methylphenylimino)diethanol, 2,2′C-(p-Tolylimino)diethanol | |
| CC1=CC=C(C=C1)N(CCO)CCO | |
| 195.26 | |
| 195.26 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-0233
TSCA : Yes