Please login to your online account to display your discounted pricing

p-Tolylacetic Acid 99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  139120250

 View more versions of this product

Catalog No. AC139120250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Fine Crystalline Powder
Formula Weight 150.18
Quantity 25g
Color White
Beilstein 09, 530
EINECS Number 210-738-7
Melting Point 90° to 93°C
RTECS Number AJ7569000
Infrared Spectrum Authentic
Molecular Formula C9H10O2
Linear Formula CH3C6H4CH2CO2H
Packaging Glass bottle
MDL Number MFCD00004353
Synonym 4-Methylphenylacetic acid
Health Hazard 1 Warning
Health Hazard 2 GHS P Statement Causes serious eye irritation. Causes skin irritation. May cause respiratory irritation.
Health Hazard 3 GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously
Chemical Name or Material p-Tolylacetic acid
Name Note 99%
Assay Percent Range 99%
CAS 622-47-9

Specifications

Chemical Name or Material p-Tolylacetic acid
Name Note 99%
CAS 622-47-9
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H10O2
Linear Formula CH3C6H4CH2CO2H
Formula Weight 150.18
MDL Number MFCD00004353
Synonym 4-Methylphenylacetic acid
Accessories

Warning

GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously

GHS P Statement Causes serious eye irritation. Causes skin irritation. May cause respiratory irritation.

SDS & Safety Documents