missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ p-Tolylacetic acid, 99%

Manufacturer:  Alfa Aesar™ A1512922

 View more versions of this product

Catalog No. AAA1512922



Specifications

Specifications

p-Tolylacetic acid
90°C to 93°C
622-47-9
MFCD00004353
p-tolylacetic acid, 4-methylphenylacetic acid, 2-p-tolyl acetic acid, 2-4-methylphenyl acetic acid, p-methylphenylacetic acid, 4-tolylacetic acid, 4-methylphenyl acetic acid, benzeneacetic acid, 4-methyl, acetic acid, p-tolyl, 2-p-tolylacetic acid
CC1=CC=C(C=C1)CC(=O)O
150.177
150.18
265°C to 267°C
100g
C9H10O2
2043528
GXXXUZIRGXYDFP-UHFFFAOYSA-N
2-(4-methylphenyl)acetic acid
248474
99%

Chemical Identifiers

622-47-9
150.177
GXXXUZIRGXYDFP-UHFFFAOYSA-N
248474
CC1=CC=C(C=C1)CC(=O)O
C9H10O2
MFCD00004353
p-tolylacetic acid, 4-methylphenylacetic acid, 2-p-tolyl acetic acid, 2-4-methylphenyl acetic acid, p-methylphenylacetic acid, 4-tolylacetic acid, 4-methylphenyl acetic acid, benzeneacetic acid, 4-methyl, acetic acid, p-tolyl, 2-p-tolylacetic acid
2-(4-methylphenyl)acetic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : AJ7569000

Documents