Please login to your online account to display your discounted pricing

Alfa Aesar™ p-Tolylacetic acid, 99%

$27.27 - $275.74

Chemical Identifiers

Chemical Name or Material p-Tolylacetic acid
CAS 622-47-9
Molecular Formula C9H10O2
Formula Weight 150.18
MDL Number MFCD00004353
Beilstein 2043528
Synonym 4-Methylphenylacetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1512906

  SDS
alfa aesar™
A1512906
5g Each for $27.27

AAA1512922

  SDS
alfa aesar™
A1512922
100g Each for $275.74
Description & Specifications

Specifications

Formula Weight 150.18
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 2043528
Molecular Formula C9H10O2
Melting Point 90° to 93°C
Assay 99%
Quantity 5g
Chemical Name or Material p-Tolylacetic acid
MDL Number MFCD00004353
Boiling Point 265° to 267°C
CAS 622-47-9
Synonym 4-Methylphenylacetic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
RTECS Number AJ7569000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.