Learn More
p-Tolylacetic acid, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1512906
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
p-Tolylacetic acid | |
90°C to 93°C | |
C9H10O2 | |
5 g | |
p-tolylacetic acid, 4-methylphenylacetic acid, 2-p-tolyl acetic acid, 2-4-methylphenyl acetic acid, p-methylphenylacetic acid, 4-tolylacetic acid, 4-methylphenyl acetic acid, benzeneacetic acid, 4-methyl, acetic acid, p-tolyl, 2-p-tolylacetic acid | |
CC1=CC=C(C=C1)CC(=O)O | |
150.177 | |
150.18 |
622-47-9 | |
265°C to 267°C | |
MFCD00004353 | |
2043528 | |
GXXXUZIRGXYDFP-UHFFFAOYSA-N | |
2-(4-methylphenyl)acetic acid | |
248474 | |
99% |
Chemical Identifiers
622-47-9 | |
150.177 | |
GXXXUZIRGXYDFP-UHFFFAOYSA-N | |
248474 | |
CC1=CC=C(C=C1)CC(=O)O |
C9H10O2 | |
MFCD00004353 | |
p-tolylacetic acid, 4-methylphenylacetic acid, 2-p-tolyl acetic acid, 2-4-methylphenyl acetic acid, p-methylphenylacetic acid, 4-tolylacetic acid, 4-methylphenyl acetic acid, benzeneacetic acid, 4-methyl, acetic acid, p-tolyl, 2-p-tolylacetic acid | |
2-(4-methylphenyl)acetic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335-H500
EINECSNumber : 210-738-7
RTECSNumber : AJ7569000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only