Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
missing translation for 'pleaseLogIn'

Alfa Aesar™ p-Tolyl isothiocyanate, 97%

Manufacturer:  Alfa Aesar™ A1149906

 View more versions of this product

Catalog No. AAA1149906


 

Description

Description

Applications
p-Tolyl isothiocyanate has been used in the preparation of 6-[1-amino-3-(p-tolyl)-thiourea]-2-ethylbenzo[de]isoquinoline-1,3-dione. It is used as capping agent to cap the N-terminal ends of polypeptide. It is also used as pharmaceutical intermediate.

Solubility
Decompose in water.

Notes
Moisture Sensitive. Protect from humidity and water. Store under dry inert gas away from oxidizing agents and moisture.
Specifications

Specifications

p-Tolyl isothiocyanate
Pungent
23°C to 26°C
622-59-3
C8H7NS
UN2923
4-Methylphenyl isothiocyanate
CC1=CC=C(C=C1)N=C=S
149.211
149.22
107°C (224°F)
236°C to 238°C
5g
97%
MFCD00004813
386032
ABQKHKWXTUVKGF-UHFFFAOYSA-N
1-isothiocyanato-4-methylbenzene
12149

Chemical Identifiers

622-59-3
149.211
ABQKHKWXTUVKGF-UHFFFAOYSA-N
12149
C8H7NS
MFCD00004813
4-Methylphenyl isothiocyanate
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

RTECSNumber : NX9500000

SDS missing translation for 'documents'