Learn More
p-Tolyl isocyanate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1357722
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
p-Tolyl isocyanate | |
1.059 | |
66°C (150°F) | |
C8H7NO | |
MFCD00002029 | |
UN2206 | |
Moisture sensitive; May form a precipitate | |
MGYGFNQQGAQEON-UHFFFAOYSA-N | |
1-isocyanato-4-methylbenzene | |
69325 | |
133.15 |
622-58-2 | |
70°C to 72°C (10 mmHg) | |
Pungent | |
1.532 | |
100 g | |
471494 | |
p-tolyl isocyanate, 4-tolyl isocyanate, 4-methylphenyl isocyanate, p-isocyanatotoluene, 4-isocyanatotoluene, p-toluene isocyanate, p-methylphenyl isocyanate, benzene, 1-isocyanato-4-methyl, p-tolylisocyanate, 1-isocyanato-4-methyl-benzene | |
CC1=CC=C(C=C1)N=C=O | |
133.15 | |
CHEBI:53732 | |
99% |
Chemical Identifiers
622-58-2 | |
133.15 | |
MGYGFNQQGAQEON-UHFFFAOYSA-N | |
69325 | |
1-isocyanato-4-methylbenzene |
C8H7NO | |
MFCD00002029 | |
p-tolyl isocyanate, 4-tolyl isocyanate, 4-methylphenyl isocyanate, p-isocyanatotoluene, 4-isocyanatotoluene, p-toluene isocyanate, p-methylphenyl isocyanate, benzene, 1-isocyanato-4-methyl, p-tolylisocyanate, 1-isocyanato-4-methyl-benzene | |
CHEBI:53732 | |
CC1=CC=C(C=C1)N=C=O |
Safety and Handling
GHS H Statement
H334-H302-H312-H332-H335-H227-H315-H319
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
P210-P235-P260-P264b-P270-P271-P272-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
H227-H302-H314-H317-H330-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: I
EINECSNumber : 210-743-4
RTECSNumber : DA3681666
TSCA : Yes
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only