Please login to your online account to display your discounted pricing

p-Tolyl Disulfide 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  139130250

 View more versions of this product

Catalog No. AC139130250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Solid
Formula Weight 246.38
Quantity 25g
Color Colorless to light yellow
TSCA TSCA
RTECS Number JO1526250
EINECS Number 203-087-5
Melting Point 43° to 46°C
Beilstein 06, 425
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 1 GHS Signal Word: Warning
MDL Number MFCD00008547
Linear Formula (CH3C6H4)2S2
Assay Percent Range 98%
CAS 103-19-5
Chemical Name or Material p-Tolyl disulfide
Name Note 98%
Molecular Formula C14H14S2

Specifications

Chemical Name or Material p-Tolyl disulfide
Name Note 98%
CAS 103-19-5
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C14H14S2
Linear Formula (CH3C6H4)2S2
Formula Weight 246.38
MDL Number MFCD00008547
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents