Please login to your online account to display your discounted pricing

p-Tolyl Disulfide 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material p-Tolyl disulfide
Name Note 98%
CAS 103-19-5
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C14H14S2
Linear Formula (CH3C6H4)2S2
Formula Weight 246.38
MDL Number MFCD00008547
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC139130250

  SDS
thermo fisher scientific
139130250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Solid
Formula Weight 246.38
Quantity 25g
Color Colorless to light yellow
EINECS Number 203-087-5
TSCA TSCA
Melting Point 43° to 46°C
RTECS Number JO1526250
Beilstein 06, 425
Chemical Name or Material p-Tolyl disulfide
Name Note 98%
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C14H14S2
Linear Formula (CH3C6H4)2S2
Packaging Glass bottle
MDL Number MFCD00008547
CAS 103-19-5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.