missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Tolyl Crotonate, TCI America™

$71.81 - $556.13

Chemical Identifiers

CAS 41873-74-9
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.215
MDL Number MFCD00059385
InChI Key FGJQFRNEXKYBQC-ONEGZZNKSA-N
Synonym p-Cresyl Crotonate, Crotonic Acid p-Tolyl Ester, Crotonic Acid p-Cresyl Ester
PubChem CID 5352024
IUPAC Name (4-methylphenyl) (E)-but-2-enoate
SMILES CC=CC(=O)OC1=CC=C(C=C1)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C086625ML View Documents TCI America
C086625ML
25mL Each for $71.81
 
C0866500ML View Documents TCI America
C0866500ML
500mL Each for $556.13
 
Specifications

Chemical Identifiers

41873-74-9
176.215
FGJQFRNEXKYBQC-ONEGZZNKSA-N
5352024
CC=CC(=O)OC1=CC=C(C=C1)C
C11H12O2
MFCD00059385
p-Cresyl Crotonate, Crotonic Acid p-Tolyl Ester, Crotonic Acid p-Cresyl Ester
(4-methylphenyl) (E)-but-2-enoate

Specifications

158°C
25mL
C11H12O2
p-Cresyl Crotonate, Crotonic Acid p-Tolyl Ester, Crotonic Acid p-Cresyl Ester
CC=CC(=O)OC1=CC=C(C=C1)C
176.215
176.22
≥55.0% (GC)
Yellow
41873-74-9
MFCD00059385
FGJQFRNEXKYBQC-ONEGZZNKSA-N
(4-methylphenyl) (E)-but-2-enoate
5352024
Clear Liquid at 20°C
p-Tolyl Crotonate
Safety and Handling

Safety and Handling

TSCA : No