missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Toluquinone 99.0+%, TCI America™

$59.25 - $171.17

Chemical Identifiers

CAS 553-97-9
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.123
MDL Number MFCD00001603
InChI Key VTWDKFNVVLAELH-UHFFFAOYSA-N
Synonym 2-methyl-1,4-benzoquinone
PubChem CID 11122
IUPAC Name 2-methylcyclohexa-2,5-diene-1,4-dione
SMILES CC1=CC(=O)C=CC1=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M01595G View Documents Tci America
M01595G
5g Each for $59.25
 
M015925G View Documents Tci America
M015925G
25g Each for $171.17
 
Specifications

Chemical Identifiers

553-97-9
122.123
VTWDKFNVVLAELH-UHFFFAOYSA-N
11122
CC1=CC(=O)C=CC1=O
C7H6O2
MFCD00001603
2-methyl-1,4-benzoquinone
2-methylcyclohexa-2,5-diene-1,4-dione

Specifications

Yellow
5g
C7H6O2
2-methyl-1,4-benzoquinone
CC1=CC(=O)C=CC1=O
122.123
122.12
≥99.0% (GC,T)
69°C
553-97-9
MFCD00001603
VTWDKFNVVLAELH-UHFFFAOYSA-N
2-methylcyclohexa-2,5-diene-1,4-dione
11122
Crystal-Powder at 20°C
p-Toluquinone
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1002

RTECSNumber : DK6300000

TSCA : Yes