missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
p-Toluoylacetonitrile, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1095006
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
p-Toluoylacetonitrile | |
99°C to 104°C | |
MFCD00067895 | |
742923 | |
AIECDYDQPCANJK-UHFFFAOYSA-N | |
3-(4-methylphenyl)-3-oxopropanenitrile | |
522513 | |
97% |
7391-28-8 | |
C10H9NO | |
5 g | |
4-methylbenzoylacetonitrile, 4-toluoylacetonitrile, 3-4-methylphenyl-3-oxopropanenitrile, p-toluoylacetonitrile, 3-oxo-3-p-tolyl propanenitrile, 3-oxo-3-p-tolylpropanenitrile, acmc-20amdj, pubchem12072, maybridge1_004714, 4-methyl-benzoylacetonitrile | |
CC1=CC=C(C=C1)C(=O)CC#N | |
159.19 | |
159.19 |
Chemical Identifiers
7391-28-8 | |
159.19 | |
AIECDYDQPCANJK-UHFFFAOYSA-N | |
522513 | |
CC1=CC=C(C=C1)C(=O)CC#N |
C10H9NO | |
MFCD00067895 | |
4-methylbenzoylacetonitrile, 4-toluoylacetonitrile, 3-4-methylphenyl-3-oxopropanenitrile, p-toluoylacetonitrile, 3-oxo-3-p-tolyl propanenitrile, 3-oxo-3-p-tolylpropanenitrile, acmc-20amdj, pubchem12072, maybridge1_004714, 4-methyl-benzoylacetonitrile | |
3-(4-methylphenyl)-3-oxopropanenitrile |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only