Learn More
p-Toluoyl chloride, 99%, Thermo Scientific Chemicals
$58.51 - $819.74
Chemical Identifiers
CAS | 874-60-2 |
---|---|
Molecular Formula | C8H7ClO |
Molecular Weight (g/mol) | 154.593 |
MDL Number | MFCD00000696 |
InChI Key | NQUVCRCCRXRJCK-UHFFFAOYSA-N |
Synonym | p-toluoyl chloride, benzoyl chloride, 4-methyl, p-methylbenzoyl chloride, 4-toluoyl chloride, p-toluyl chloride, p-toluic acid chloride, 4-methylbenzoic acid chloride, 4-methylbenzoylchloride, 4-methyl-benzoyl chloride, unii-092e36pwl4 |
PubChem CID | 13405 |
IUPAC Name | 4-methylbenzoyl chloride |
SMILES | CC1=CC=C(C=C1)C(=O)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1558122
|
Thermo Scientific Chemicals
A1558122 |
100 g |
Each for $58.51
|
|
|||||
AAA1558136
|
Thermo Scientific Chemicals
A1558136 |
500 g |
Each for $197.62
|
|
|||||
AAA155810E
|
Thermo Scientific Chemicals
A155810E |
2500 g |
Each for $819.74
|
|
|||||
Description
p-Toluoyl chloride was used to prepare antigen, Tolyl-amido-human serum albumin. It is used as a reagent to synthesize (+)-Yashabushitriol, derivatives of Desloratadine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsp-Toluoyl chloride was used to prepare antigen, Tolyl-amido-human serum albumin. It is used as a reagent to synthesize (+)-Yashabushitriol, derivatives of Desloratadine.
Solubility
Reacts with water alcohol.
Notes
Store at room temperature. Incompatible with oxidizing agent, acids, bases, active metals, alcohols, amines.
Chemical Identifiers
874-60-2 | |
154.593 | |
NQUVCRCCRXRJCK-UHFFFAOYSA-N | |
13405 | |
CC1=CC=C(C=C1)C(=O)Cl |
C8H7ClO | |
MFCD00000696 | |
p-toluoyl chloride, benzoyl chloride, 4-methyl, p-methylbenzoyl chloride, 4-toluoyl chloride, p-toluyl chloride, p-toluic acid chloride, 4-methylbenzoic acid chloride, 4-methylbenzoylchloride, 4-methyl-benzoyl chloride, unii-092e36pwl4 | |
4-methylbenzoyl chloride |
Specifications
874-60-2 | |
1.169 | |
82°C (179°F) | |
C8H7ClO | |
MFCD00000696 | |
UN3265 | |
Moisture sensitive | |
Reacts with water alcohol. | |
CC1=CC=C(C=C1)C(=O)Cl | |
154.593 | |
154.6 | |
Liquid |
°C to 2°C | |
225°C to 227°C | |
99% | |
1.553 | |
100 g | |
471492 | |
p-toluoyl chloride, benzoyl chloride, 4-methyl, p-methylbenzoyl chloride, 4-toluoyl chloride, p-toluyl chloride, p-toluic acid chloride, 4-methylbenzoic acid chloride, 4-methylbenzoylchloride, 4-methyl-benzoyl chloride, unii-092e36pwl4 | |
NQUVCRCCRXRJCK-UHFFFAOYSA-N | |
4-methylbenzoyl chloride | |
13405 | |
99% | |
p-Toluoyl Chloride |
Safety and Handling
GHS H Statement
H314-H318-H227
P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 212-864-8
RTECSNumber : XV1660000
TSCA : Yes
storageNote1 : Moisture sensitive
Recommended Storage : Ambient temperatures
RUO – Research Use Only