missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Toluoyl chloride, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1558136

 View more versions of this product

Catalog No. AAA1558136


Description

Description

p-Toluoyl chloride was used to prepare antigen, Tolyl-amido-human serum albumin. It is used as a reagent to synthesize (+)-Yashabushitriol, derivatives of Desloratadine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Toluoyl chloride was used to prepare antigen, Tolyl-amido-human serum albumin. It is used as a reagent to synthesize (+)-Yashabushitriol, derivatives of Desloratadine.

Solubility
Reacts with water alcohol.

Notes
Store at room temperature. Incompatible with oxidizing agent, acids, bases, active metals, alcohols, amines.
Specifications

Specifications

p-Toluoyl chloride
-2°C
225°C to 227°C
99%
C8H7ClO
MFCD00000696
UN3265
Moisture sensitive
Reacts with water alcohol.
CC1=CC=C(C=C1)C(=O)Cl
154.593
154.6
Liquid
874-60-2
1.169
82°C (179°F)
Pungent
1.553
500 g
471492
p-toluoyl chloride, benzoyl chloride, 4-methyl, p-methylbenzoyl chloride, 4-toluoyl chloride, p-toluyl chloride, p-toluic acid chloride, 4-methylbenzoic acid chloride, 4-methylbenzoylchloride, 4-methyl-benzoyl chloride, unii-092e36pwl4
NQUVCRCCRXRJCK-UHFFFAOYSA-N
4-methylbenzoyl chloride
13405
99%

Chemical Identifiers

874-60-2
154.593
NQUVCRCCRXRJCK-UHFFFAOYSA-N
13405
CC1=CC=C(C=C1)C(=O)Cl
C8H7ClO
MFCD00000696
p-toluoyl chloride, benzoyl chloride, 4-methyl, p-methylbenzoyl chloride, 4-toluoyl chloride, p-toluyl chloride, p-toluic acid chloride, 4-methylbenzoic acid chloride, 4-methylbenzoylchloride, 4-methyl-benzoyl chloride, unii-092e36pwl4
4-methylbenzoyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 212-864-8

RTECSNumber : XV1660000

TSCA : Yes

storageNote1 : Moisture sensitive

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only