Learn More
p-Toluoyl chloride, 98%, Thermo Scientific Chemicals
$46.20 - $107.04
Chemical Identifiers
CAS | 874-60-2 |
---|---|
Molecular Formula | C8H7ClO |
Molecular Weight (g/mol) | 154.6 |
MDL Number | MFCD00000696 |
InChI Key | NQUVCRCCRXRJCK-UHFFFAOYSA-N |
Synonym | p-toluoyl chloride, benzoyl chloride, 4-methyl, p-methylbenzoyl chloride, 4-toluoyl chloride, p-toluyl chloride, p-toluic acid chloride, 4-methylbenzoic acid chloride, 4-methylbenzoylchloride, 4-methyl-benzoyl chloride, unii-092e36pwl4 |
PubChem CID | 13405 |
IUPAC Name | 4-methylbenzoyl chloride |
SMILES | CC1=CC=C(C=C1)C(=O)Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC147980050
|
Thermo Scientific Chemicals
147980050 |
5 mL | Glass bottle |
Each for $46.20
|
|
||||
AC147982500
|
Thermo Scientific Chemicals
147982500 |
250 mL | Glass bottle |
Each for $107.04
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
874-60-2 | |
154.6 | |
NQUVCRCCRXRJCK-UHFFFAOYSA-N | |
13405 | |
CC1=CC=C(C=C1)C(=O)Cl |
C8H7ClO | |
MFCD00000696 | |
p-toluoyl chloride, benzoyl chloride, 4-methyl, p-methylbenzoyl chloride, 4-toluoyl chloride, p-toluyl chloride, p-toluic acid chloride, 4-methylbenzoic acid chloride, 4-methylbenzoylchloride, 4-methyl-benzoyl chloride, unii-092e36pwl4 | |
4-methylbenzoyl chloride |
Specifications
874-60-2 | |
Colorless to Brown | |
225.0°C to 227.0°C | |
Authentic | |
Glass bottle | |
1.5525 to 1.5545 | |
MFCD00000696 | |
09,486 | |
p-toluoyl chloride, benzoyl chloride, 4-methyl, p-methylbenzoyl chloride, 4-toluoyl chloride, p-toluyl chloride, p-toluic acid chloride, 4-methylbenzoic acid chloride, 4-methylbenzoylchloride, 4-methyl-benzoyl chloride, unii-092e36pwl4 | |
NQUVCRCCRXRJCK-UHFFFAOYSA-N | |
4-methylbenzoyl chloride | |
13405 | |
98% | |
p-Toluoyl chloride, 98% |
-2.0°C | |
1.1600g/mL | |
82°C | |
98% | |
C8H7ClO | |
CH3C6H4COCl | |
5 mL | |
1.16 | |
Solubility in water: decomposes. | |
CC1=CC=C(C=C1)C(=O)Cl | |
154.6 | |
154.6 | |
Liquid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 212-864-8
RTECSNumber : XV1660000
TSCA : TSCA
RUO – Research Use Only