missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Tolunitrile, 98+%, ACROS Organics™

Manufacturer:  Acros Organics 158022500

 View more versions of this product

Catalog No. AC158022500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

p-Tolunitrile
85°C
Glass bottle
0.98
103.0°C to 106.0°C (20.0 mmHg)
26.0°C to 28.0°C
98%
98+%
CH3C6H4CN
09, 489
p-tolunitrile, 4-cyanotoluene, benzonitrile, 4-methyl, 4-tolunitrile, p-methylbenzonitrile, p-toluonitrile, p-cyanotoluene, p-tolylnitrile, p-tolunitril, p-toluenenitrile
VCZNNAKNUVJVGX-UHFFFAOYSA-N
4-methylbenzonitrile
7724
Viscous Liquid
0.9800g/mL
Authentic
1.5285 to 1.5305
0.5% max. (K.F.)
Undesignated
250g
104-85-8
C8H7N
MFCD00001827
15, 9698
Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
CC1=CC=C(C=C1)C#N
117.151
117.15
≥98%

Chemical Identifiers

104-85-8
117.151
VCZNNAKNUVJVGX-UHFFFAOYSA-N
7724
CC1=CC=C(C=C1)C#N
C8H7N
MFCD00001827
p-tolunitrile, 4-cyanotoluene, benzonitrile, 4-methyl, 4-tolunitrile, p-methylbenzonitrile, p-toluonitrile, p-cyanotoluene, p-tolylnitrile, p-tolunitril, p-toluenenitrile
4-methylbenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 203-244-8

RTECSNumber : XV0700000

TSCA : TSCA

SDS Documents