Please login to your online account to display your discounted pricing

p-Toluic Hydrazide 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  139070050

 View more versions of this product

Catalog No. AC139070050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 150.18
Physical Form Crystalline Powder
Quantity 5g
Color White to slightly brown
EINECS Number 222-815-2
Melting Point 114° to 118°C
Beilstein 09, 494
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material p-Toluic hydrazide
Name Note 98%
Assay Percent Range 98%
Molecular Formula C8H10N2O
Linear Formula CH3C6H4CONHNH2
MDL Number MFCD00007607
CAS 3619-22-5

Specifications

Chemical Name or Material p-Toluic hydrazide
Name Note 98%
CAS 3619-22-5
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H10N2O
Linear Formula CH3C6H4CONHNH2
Formula Weight 150.18
MDL Number MFCD00007607
Accessories

GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents