Learn More
p-Toluenethiol, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 138711000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
p-Toluenethiol | |
106-45-6 | |
White to Yellow | |
68°C | |
97.5% min. (GC) | |
C7H8S | |
MFCD00004851 | |
06, 416 | |
p-toluenethiol, p-thiocresol, 4-thiocresol, 4-methylthiophenol, p-methylthiophenol, benzenethiol, 4-methyl, 4-toluenethiol, p-tolyl mercaptan, p-thiolcresol, p-tolylthiol | |
WLHCBQAPPJAULW-UHFFFAOYSA-N | |
4-methylbenzenethiol | |
7811 | |
98% |
98% | |
40.0°C to 44.0°C | |
195.0°C | |
Authentic | |
Glass bottle | |
CH3C6H4SH | |
100 g | |
15, 9478 | |
Solubility in water: slightly soluble. Other solubilities: soluble in alcohol and ether | |
CC1=CC=C(C=C1)S | |
124.2 | |
124.2 | |
Fused Solid |
Chemical Identifiers
106-45-6 | |
124.2 | |
WLHCBQAPPJAULW-UHFFFAOYSA-N | |
7811 | |
CC1=CC=C(C=C1)S |
C7H8S | |
MFCD00004851 | |
p-toluenethiol, p-thiocresol, 4-thiocresol, 4-methylthiophenol, p-methylthiophenol, benzenethiol, 4-methyl, 4-toluenethiol, p-tolyl mercaptan, p-thiolcresol, p-tolylthiol | |
4-methylbenzenethiol |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air
GHS Signal Word: Warning
EINECSNumber : 203-399-1
RTECSNumber : XT8925000
TSCA : TSCA
RUO – Research Use Only