Please login to your online account to display your discounted pricing

p-Toluenesulfonylacetonitrile, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material p-Toluenesulfonylacetonitrile
Name Note 98%
CAS 5697-44-9
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H9NO2S
Formula Weight 195.24
MDL Number MFCD00039487
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC351720250

  SDS
thermo fisher scientific
AC351720250
Glass Bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥97.5%
Formula Weight 195.24
Quantity 25g
Color Off-white
Melting Point 147° to 151°C
EINECS Number 227-173-7
Chemical Name or Material p-Toluenesulfonylacetonitrile
Name Note 98%
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
MDL Number MFCD00039487
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C9H9NO2S
CAS 5697-44-9