missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
p-Toluenesulfonyl cyanide, 95%, Thermo Scientific™
Chemical Identifiers
CAS | 19158-51-1 |
---|---|
Molecular Formula | C8H7NO2S |
Molecular Weight (g/mol) | 181.21 |
InChI Key | JONIMGVUGJVFQD-UHFFFAOYSA-N |
Synonym | tosyl cyanide, p-toluenesulfonyl cyanide, 4-methylbenzenesulfonyl cyanide, p-toluenesulphonyl cyanide, p-toluene sulfonyl cyanide, p-tolylsulfonyl cyanide, p-toluenesulfonylcyanide, benzenesulfonylcyanide, 4-methyl, benzenesulfonyl cyanide, 4-methyl, 4-methylphenyl sulfonylformonitrile |
PubChem CID | 87946 |
IUPAC Name | (4-methylphenyl)sulfonylformonitrile |
SMILES | CC1=CC=C(C=C1)S(=O)(=O)C#N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC455820010
|
Thermo Scientific Chemicals
455820010 |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC455820050
|
Thermo Scientific Chemicals
455820050 |
5g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
19158-51-1 | |
181.21 | |
tosyl cyanide, p-toluenesulfonyl cyanide, 4-methylbenzenesulfonyl cyanide, p-toluenesulphonyl cyanide, p-toluene sulfonyl cyanide, p-tolylsulfonyl cyanide, p-toluenesulfonylcyanide, benzenesulfonylcyanide, 4-methyl, benzenesulfonyl cyanide, 4-methyl, 4-methylphenyl sulfonylformonitrile | |
(4-methylphenyl)sulfonylformonitrile |
C8H7NO2S | |
JONIMGVUGJVFQD-UHFFFAOYSA-N | |
87946 | |
CC1=CC=C(C=C1)S(=O)(=O)C#N |
Specifications
19158-51-1 | |
94% min. (HPLC) | |
C8H7NO2S | |
tosyl cyanide, p-toluenesulfonyl cyanide, 4-methylbenzenesulfonyl cyanide, p-toluenesulphonyl cyanide, p-toluene sulfonyl cyanide, p-tolylsulfonyl cyanide, p-toluenesulfonylcyanide, benzenesulfonylcyanide, 4-methyl, benzenesulfonyl cyanide, 4-methyl, 4-methylphenyl sulfonylformonitrile | |
CC1=CC=C(C=C1)S(=O)(=O)C#N | |
181.21 | |
181.21 | |
p-Toluenesulfonyl cyanide |
Authentic | |
Glass Bottle | |
1g | |
JONIMGVUGJVFQD-UHFFFAOYSA-N | |
(4-methylphenyl)sulfonylformonitrile | |
87946 | |
95% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective
GHS Signal Word: Danger
EINECSNumber : 242-849-1
RUO â Research Use Only