Learn More
p-Toluenesulfonyl chloride, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 139030100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
p-Toluenesulfonyl chloride | |
67.0°C to 69.0°C | |
134.0°C (10.0 mmHg) | |
Authentic | |
Plastic drum | |
CH3C6H4SO2Cl | |
10 kg | |
01,1179; 03,292; 04,510; 05,676; 06,598; 08,489; 11,536; 13,286 | |
tosyl chloride, p-toluenesulfonyl chloride, p-tosyl chloride, 4-toluenesulfonyl chloride, p-toluenesulfochloride, 4-methylbenzene-1-sulfonyl chloride, p-tolylsulfonyl chloride, p-toluenesulphonyl chloride, tosylchloride, p-toluenesulfonic acid chloride | |
YYROPELSRYBVMQ-UHFFFAOYSA-N | |
4-methylbenzenesulfonyl chloride | |
7397 | |
99+% |
98-59-9 | |
White | |
128°C | |
99% min | |
C7H7ClO2S | |
MFCD00007450 | |
11,103 | |
15,9693 | |
Solubility in water: hydrolyses. Other solubilities: hydrolysis to acid, freely soluble in alcohol, benzene and ether | |
CC1=CC=C(C=C1)S(Cl)(=O)=O | |
190.64 | |
190.65 | |
Crystalline Powder |
Chemical Identifiers
98-59-9 | |
190.64 | |
YYROPELSRYBVMQ-UHFFFAOYSA-N | |
7397 | |
CC1=CC=C(C=C1)S(Cl)(=O)=O |
C7H7ClO2S | |
MFCD00007450 | |
tosyl chloride, p-toluenesulfonyl chloride, p-tosyl chloride, 4-toluenesulfonyl chloride, p-toluenesulfochloride, 4-methylbenzene-1-sulfonyl chloride, p-tolylsulfonyl chloride, p-toluenesulphonyl chloride, tosylchloride, p-toluenesulfonic acid chloride | |
4-methylbenzenesulfonyl chloride |
Safety and Handling
GHS H Statement
May be corrosive to metals.
Causes skin irritation.
Causes serious eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 202-684-8
RUO – Research Use Only