missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Toluenesulfonic Anhydride, TCI America™

Manufacturer:  TCI America T14855G

 View more versions of this product

Catalog No. T14855G



Specifications

Specifications

p-Toluenesulfonic Anhydride
125°C
4124-41-8
MFCD00008548
p-toluenesulfonic anhydride, 4-methylbenzenesulfonic anhydride, 4-toluenesulfonic anhydride, p-toluenesulfonicanhydride, p-toluene sulfonic anhydride, toluene-p-sulphonic anhydride, 4-methylphenyl sulfonyl 4-methylbenzenesulfonate, p-toluenesulphonic anhydride, toluene-4-sulfonic anhydride, benzenesulfonic acid, 4-methyl-, anhydride
CC1=CC=C(C=C1)S(=O)(=O)OS(=O)(=O)C2=CC=C(C=C2)C
326.381
326.38
≥95.0% (T)
White
5g
C14H14O5S2
1759
PDVFSPNIEOYOQL-UHFFFAOYSA-N
(4-methylphenyl)sulfonyl 4-methylbenzenesulfonate
77773
Crystal-Powder at 20°C

Chemical Identifiers

4124-41-8
326.381
PDVFSPNIEOYOQL-UHFFFAOYSA-N
77773
CC1=CC=C(C=C1)S(=O)(=O)OS(=O)(=O)C2=CC=C(C=C2)C
C14H14O5S2
MFCD00008548
p-toluenesulfonic anhydride, 4-methylbenzenesulfonic anhydride, 4-toluenesulfonic anhydride, p-toluenesulfonicanhydride, p-toluene sulfonic anhydride, toluene-p-sulphonic anhydride, 4-methylphenyl sulfonyl 4-methylbenzenesulfonate, p-toluenesulphonic anhydride, toluene-4-sulfonic anhydride, benzenesulfonic acid, 4-methyl-, anhydride
(4-methylphenyl)sulfonyl 4-methylbenzenesulfonate
Safety and Handling

Safety and Handling

TSCA : No

Documents