Please login to your online account to display your discounted pricing

p-Toluenesulfonic Acid, Sodium Salt, Mixture of Isomers 99%, ACROS Organics™


Quantity 100g

Manufacturer: thermo fisher scientific  259471000

 View more versions of this product

Catalog No. AC259471000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Fine Crystalline Powder
Formula Weight 194.19
Quantity 100g
Color White
RTECS Number XT7350000
EINECS Number 211-522-5
TSCA TSCA
pH 7 to 10 (1% aq. soln.)
Beilstein 11, 97
Fieser 11,535; 12,507
Merck Index 14, 9533
Synonym Sodium p-toluenesulfonate
Infrared Spectrum Authentic
MDL Number MFCD00064388
Name Note 99%
Molecular Formula C7H7NaO3S
CAS 657-84-1
Chemical Name or Material p-Toluenesulfonic acid, sodium salt, mixture of isomers
Linear Formula CH3C6H4SO3Na

Specifications

Chemical Name or Material p-Toluenesulfonic acid, sodium salt, mixture of isomers
Name Note 99%
CAS 657-84-1
Assay ≥98.5%
Molecular Formula C7H7NaO3S
Linear Formula CH3C6H4SO3Na
Formula Weight 194.19
MDL Number MFCD00064388
Synonym Sodium p-toluenesulfonate