Please login to your online account to display your discounted pricing

p-Toluenesulfonic acid, sodium salt, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: fisher scientific acros  AC421231000

 View more versions of this product

Catalog No. AC421231000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5% (ion exchange; alkalimetry)
Physical Form Powder
Formula Weight 194.19
Quantity 100g
Color White to off-white
Packaging Glass bottle
Synonym Sodium p-toluenesulfonate
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
MDL Number MFCD00064388
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 657-84-1
Molecular Formula C7H7NaO3S
Chemical Name or Material p-Toluenesulfonic acid, sodium salt

Specifications

Chemical Name or Material p-Toluenesulfonic acid, sodium salt
Assay ≥97.5% (ion exchange; alkalimetry)
Molecular Formula C7H7NaO3S
Formula Weight 194.19
MDL Number MFCD00064388
Synonym Sodium p-toluenesulfonate
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents