Please login to your online account to display your discounted pricing

Alfa Aesar™ p-Toluenesulfonic acid, polymer-supported, 1.0-2.0 mmol/g on polystyrene

Chemical Identifiers

Chemical Name or Material p-Toluenesulfonic acid
Name Note 1.0 to 2.0mmol/g on polystyrene
MDL Number MFCD02683442
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1975506

  SDS
alfa aesar™
L1975506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1975514

  SDS
alfa aesar™
L1975514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Name Note 1.0 to 2.0mmol/g on polystyrene
Quantity 5g
MDL Number MFCD02683442
Chemical Name or Material p-Toluenesulfonic acid