Learn More
p-Toluenesulfonic acid monohydrate, 97.5%, pure, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 171780050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
p-Toluenesulfonic acid monohydrate | |
180°C | |
97.5% | |
C7H10O4S | |
MFCD00142137 | |
11, 97 | |
15, 9692 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and chloroform | |
CC1=CC=C(C=C1)S(=O)(=O)O.O | |
190.213 | |
190.22 | |
Pure |
6192-52-5 | |
Authentic | |
Plastic drum | |
CH3C6H4SO3H·H2O | |
5kg | |
01,1172; 04,508; 05,673; 06,597; 07,374; 08,488; 09,471; 11,535; 12,507 | |
p-toluenesulfonic acid monohydrate, 4-methylbenzenesulfonic acid hydrate, 4-methylbenzenesulfonic acid monohydrate, p-toluene sulfonic acid monohydrate, unii-3bto78gaff, benzenesulfonic acid, 4-methyl-, monohydrate, ptoluenesulfonic acid, p-toluenesulfonic acid, monohydrate, 3bto78gaff, 4-methylbenzene-1-sulfonic acid hydrate | |
KJIFKLIQANRMOU-UHFFFAOYSA-N | |
4-methylbenzenesulfonic acid;hydrate | |
521998 | |
97.5% |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
Recommended Storage : Product becomes pink on ageing
RUO â Research Use Only