missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ p-Toluenesulfonanilide, 97%

Manufacturer:  Alfa Aesar™ H5556403

 View more versions of this product

Catalog No. AAH5556403



Specifications

Specifications

p-Toluenesulfonanilide
1g
C13H13NO2S
4-methyl-n-phenyl-benzenesulfonamide
CC1=CC=C(C=C1)S(=O)(=O)NC2=CC=CC=C2
247.312
247.31
102°C
68-34-8
MFCD00025989
VLVCWODDMDGANW-UHFFFAOYSA-N
4-methyl-N-phenylbenzenesulfonamide
2456
97%

Chemical Identifiers

68-34-8
247.312
VLVCWODDMDGANW-UHFFFAOYSA-N
2456
CC1=CC=C(C=C1)S(=O)(=O)NC2=CC=CC=C2
C13H13NO2S
MFCD00025989
4-methyl-n-phenyl-benzenesulfonamide
4-methyl-N-phenylbenzenesulfonamide
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : XT6100000

TSCA : TSCA

Documents