Learn More
p-Toluenesulfinic acid, sodium salt hydrate, 98+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 139011000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
p-Toluenesulfinic acid,sodium salt hydrate | |
207801-20-5 | |
98+% | |
C7H7NaO2S·xH2O | |
MFCD00149640 | |
11, 9 | |
10, 9360 | |
Solubility in water: soluble | |
CC1=CC=C(C=C1)S(=O)[O-].O.[Na+] | |
178.18 | |
178.18 |
98+% | |
Authentic | |
Plastic bottle | |
CH3C6H4SO2Na·xH2O | |
100g | |
08,488 | |
sodium p-toluenesulfinate hydrate, benzenesulfinic acid, 4-methyl-, sodium salt, hydrate, c7h7o2s.na.h2o, p-toluenesulfinic acid sodium salt, sodium 4-toluenesulphinate monohydrate, p-toluenesulfinic acid sodium salt hydrate, sodium 4-methylbenzenesulfinate hydrate, p-toluenesulfinic acid sodium salt dry wt. , water | |
WUHWAOGAJFMIFU-UHFFFAOYSA-M | |
sodium;4-methylbenzenesulfinate;hydrate | |
23682957 | |
98+% |
Chemical Identifiers
207801-20-5 | |
178.18 | |
WUHWAOGAJFMIFU-UHFFFAOYSA-M | |
23682957 | |
CC1=CC=C(C=C1)S(=O)[O-].O.[Na+] |
C7H7NaO2S·xH2O | |
MFCD00149640 | |
sodium p-toluenesulfinate hydrate, benzenesulfinic acid, 4-methyl-, sodium salt, hydrate, c7h7o2s.na.h2o, p-toluenesulfinic acid sodium salt, sodium 4-toluenesulphinate monohydrate, p-toluenesulfinic acid sodium salt hydrate, sodium 4-methylbenzenesulfinate hydrate, p-toluenesulfinic acid sodium salt dry wt. , water | |
sodium;4-methylbenzenesulfinate;hydrate |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
EINECSNumber : 212-538-5
RTECSNumber : XT4725000
TSCA : TSCA
RUO â Research Use Only