missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Toluenesulfinic acid, sodium salt, 97%, anhydrous, ACROS Organics™

Manufacturer:  Acros Organics AC434531000

 View more versions of this product

Catalog No. AC434531000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

p-Toluenesulfinic acid, sodium salt
Glass bottle
White
100g
97%
sodium 4-methylbenzenesulfinate, sodium p-toluenesulfinate, p-toluenesulfinic acid sodium salt, sodium toluene-4-sulphinate, sodium p-tolylsulfinate, sodium 4-toluenesulfinate, benzenesulfinic acid, 4-methyl-, sodium salt, unii-7cku5w4trm, p-toluenesulfinic acid, sodium salt, p-toluensulfinan sodny czech
KFZUDNZQQCWGKF-UHFFFAOYSA-M
sodium;4-methylbenzenesulfinate
2723791
Powder
Authentic
3% Max.
>300.0°C
824-79-3
C7H7NaO2S
Solubility in water: soluble.
CC1=CC=C(C=C1)S(=O)[O-].[Na+]
178.181
178.18
97%

Chemical Identifiers

824-79-3
178.181
sodium 4-methylbenzenesulfinate, sodium p-toluenesulfinate, p-toluenesulfinic acid sodium salt, sodium toluene-4-sulphinate, sodium p-tolylsulfinate, sodium 4-toluenesulfinate, benzenesulfinic acid, 4-methyl-, sodium salt, unii-7cku5w4trm, p-toluenesulfinic acid, sodium salt, p-toluensulfinan sodny czech
sodium;4-methylbenzenesulfinate
C7H7NaO2S
KFZUDNZQQCWGKF-UHFFFAOYSA-M
2723791
CC1=CC=C(C=C1)S(=O)[O-].[Na+]
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents