Please login to your online account to display your discounted pricing

p-Tolualdehyde, +99%, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: fisher scientific acros  421145000

 View more versions of this product

Catalog No. AC421145000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Liquid
Formula Weight 120.15
Quantity 500g
Color Clear colorless to pale yellow
EINECS Number 203-246-9
TSCA TSCA
Specific Gravity 1.010 to 1.025 (20°C)
Density 1.015g/mL
RTECS Number CU7034500
Chemical Name or Material p-Tolualdehyde
Infrared Spectrum Authentic
Molecular Formula C8H8O
Linear Formula CH3C6H4CHO
Packaging Glass bottle
MDL Number MFCD00006954
CAS 104-87-0
Refractive Index 1.5440 to 1.5480 (20°C, 589nm)
Synonym 4-Methylbenzaldehyde
Health Hazard 1 AIR SENSITIVE
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 99+%
Assay Percent Range 99+%

Specifications

Chemical Name or Material p-Tolualdehyde
Name Note 99+%
CAS 104-87-0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C8H8O
Linear Formula CH3C6H4CHO
Formula Weight 120.15
MDL Number MFCD00006954
Synonym 4-Methylbenzaldehyde
Accessories

AIR SENSITIVE

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents