missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Quaterphenyl 98.0+%, TCI America™

Manufacturer:  Tci America Q00015G

 View more versions of this product

Catalog No. Q00015G



Specifications

Specifications

p-Quaterphenyl
5g
C24H18
p-quaterphenyl, 1,1':4',1:4,1'-quaterphenyl, benzerythrene, quadriphenyl, p-tetraphenyl, 4,4'-diphenylbiphenyl, p,p'-quaterphenyl, 4-quaterphenyl, unii-g8aqm6d0rk, 1-phenyl-4-4-phenylphenyl benzene
C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=C(C=C3)C4=CC=CC=C4
306.408
CHEBI:52240
Crystal-Powder at 20°C
White
135-70-6
MFCD00003062
GPRIERYVMZVKTC-UHFFFAOYSA-N
1-phenyl-4-(4-phenylphenyl)benzene
8677
306.41
≥98.0% (HPLC)

Chemical Identifiers

135-70-6
306.408
GPRIERYVMZVKTC-UHFFFAOYSA-N
8677
1-phenyl-4-(4-phenylphenyl)benzene
C24H18
MFCD00003062
p-quaterphenyl, 1,1':4',1:4,1'-quaterphenyl, benzerythrene, quadriphenyl, p-tetraphenyl, 4,4'-diphenylbiphenyl, p,p'-quaterphenyl, 4-quaterphenyl, unii-g8aqm6d0rk, 1-phenyl-4-4-phenylphenyl benzene
CHEBI:52240
C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=C(C=C3)C4=CC=CC=C4
Safety and Handling

Safety and Handling

TSCA : Yes

Documents