Please login to your online account to display your discounted pricing

Alfa Aesar™ p-Quaterphenyl, 99.5+%

$261.68 - $261.68

Chemical Identifiers

Chemical Name or Material p-Quaterphenyl
CAS 135-70-6
Molecular Formula C24H18
Formula Weight 306.41
MDL Number MFCD00003062
Beilstein 1912745
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1778806

 
alfa aesar™
A1778806
5g Each for $261.68
Description & Specifications

Specifications

Formula Weight 306.41
Chemical Name or Material p-Quaterphenyl
Assay ≥99.5%
Molecular Formula C24H18
MDL Number MFCD00003062
CAS 135-70-6
Melting Point 316° to 318°C
Quantity 5g
TSCA TSCA
Beilstein 1912745
Boiling Point 428°C (18mmHg)