Please login to your online account to display your discounted pricing

Alfa Aesar™ p-Phenylenediamine sulfate, 98%

Chemical Identifiers

Chemical Name or Material p-Phenylenediamine sulfate
CAS 16245-77-5
Molecular Formula C6H8N2·H2SO4
Formula Weight 206.22
MDL Number MFCD00035510
UN Number UN2811
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA126590B

  SDS
alfa aesar™
A126590B
1000g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1265930

  SDS
alfa aesar™
A1265930
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 206.22
Assay 98%
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA TSCA
Health Hazard 2 GHS H Statement
H301-H311-H331-H341-H319-H317
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Suspected of causing genetic defects.
Causes serious eye irritation.
May cause an allergic skin reaction.
Melting Point >300°C
UN Number UN2811
Chemical Name or Material p-Phenylenediamine sulfate
CAS 16245-77-5
MDL Number MFCD00035510
Health Hazard 1 Danger
Quantity 1000g
Health Hazard 3 GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C6H8N2·H2SO4
Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H331-H341-H319-H317
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Suspected of causing genetic defects.
Causes serious eye irritation.
May cause an allergic skin reaction.