Please login to your online account to display your discounted pricing

Alfa Aesar™ p-Phenylenediamine dihydrochloride, 99%

Chemical Identifiers

Chemical Name or Material p-Phenylenediamine dihydrochloride
CAS 624-18-0
Molecular Formula C6H8N2·2HCl
Formula Weight 181.07
MDL Number MFCD00013002
UN Number UN2811
Beilstein 3911749
Merck Index 14,7285
Synonym 1,4-Diaminobenzene dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA106380B

  SDS
alfa aesar™
A106380B
1000g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1063818

  SDS
alfa aesar™
A1063818
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1063830

  SDS
alfa aesar™
A1063830
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 181.07
Synonym 1,4-Diaminobenzene dihydrochloride
Health Hazard 1 Danger
Merck Index 14,7285
CAS 624-18-0
TSCA TSCA
Assay 99%
UN Number UN2811
Melting Point ≈265°C
Quantity 1000g
Molecular Formula C6H8N2·2HCl
MDL Number MFCD00013002
Health Hazard 2 GHS H Statement
H301-H311-H331-H319-H317
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes serious eye irritation.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECS Number ST0350000
Beilstein 3911749
Chemical Name or Material p-Phenylenediamine dihydrochloride
Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H331-H319-H317
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes serious eye irritation.
May cause an allergic skin reaction.