Please login to your online account to display your discounted pricing

p-Phenylene Diisothiocyanate 99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  417510250

 View more versions of this product

Catalog No. AC417510250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Crystalline Needles or Powder
Formula Weight 192.27
Quantity 25g
Color Off-white to yellow to beige-gray
EINECS Number 223-741-3
TSCA TSCA
RTECS Number NX9150000
Melting Point 129° to 133°C
Merck Index 15, 1309
Molecular Formula C8H4N2S2
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material p-Phenylene diisothiocyanate
Name Note 99%
Assay Percent Range 99%
Linear Formula SCNC6H4NCS
MDL Number MFCD00004811
CAS 4044-65-9

Specifications

Chemical Name or Material p-Phenylene diisothiocyanate
Name Note 99%
CAS 4044-65-9
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H4N2S2
Linear Formula SCNC6H4NCS
Formula Weight 192.27
MDL Number MFCD00004811
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

SDS & Safety Documents