Learn More
p-Phenylene diisothiocyanate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 417510050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
p-Phenylene diisothiocyanate | |
4044-65-9 | |
Beige-Gray to White | |
98.5% min. (GC) | |
C8H4N2S2 | |
MFCD00004811 | |
15, 1309 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol and chloroform,slightly soluble in acetone | |
C1=CC(=CC=C1N=C=S)N=C=S | |
192.27 | |
192.27 | |
Crystalline Needles or Powder |
99% | |
129.0°C to 133.0°C | |
Authentic | |
Glass bottle | |
SCNC6H4NCS | |
5 g | |
bitoscanate, 1,4-phenylene diisothiocyanate, jonit, p-phenylene diisothiocyanate, benzene, 1,4-diisothiocyanato, bitoscanat, bitoscanate inn, phenylene thiocyanate, bitoscanatum latin, 1,4-phenylenediisothiocyanate | |
OMWQUXGVXQELIX-UHFFFAOYSA-N | |
1,4-diisothiocyanatobenzene | |
19958 | |
99% |
Chemical Identifiers
4044-65-9 | |
192.27 | |
OMWQUXGVXQELIX-UHFFFAOYSA-N | |
19958 | |
C1=CC(=CC=C1N=C=S)N=C=S |
C8H4N2S2 | |
MFCD00004811 | |
bitoscanate, 1,4-phenylene diisothiocyanate, jonit, p-phenylene diisothiocyanate, benzene, 1,4-diisothiocyanato, bitoscanat, bitoscanate inn, phenylene thiocyanate, bitoscanatum latin, 1,4-phenylenediisothiocyanate | |
1,4-diisothiocyanatobenzene |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep a
GHS Signal Word: Danger
EINECSNumber : 223-741-3
RTECSNumber : NX9150000
TSCA : TSCA
RUO – Research Use Only