missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Phenetidine Hydrochloride, TCI America™

Manufacturer:  TCI America P0116500G

 View more versions of this product

Catalog No. P0116500G



Specifications

Specifications

p-Phenetidine Hydrochloride
500g
C8H12ClNO
4-Ethoxyaniline Hydrochloride
CCOC1=CC=C(C=C1)N.Cl
173.64
173.64
≥98.0% (HPLC,T)
Red-White
637-56-9
MFCD00035503
JVWYCUBGJVOEIZ-UHFFFAOYSA-N
4-ethoxyaniline;hydrochloride
69478
Crystal-Powder at 20°C

Chemical Identifiers

637-56-9
173.64
JVWYCUBGJVOEIZ-UHFFFAOYSA-N
69478
CCOC1=CC=C(C=C1)N.Cl
C8H12ClNO
MFCD00035503
4-Ethoxyaniline Hydrochloride
4-ethoxyaniline;hydrochloride
Safety and Handling

Safety and Handling

RTECSNumber : SI7051000

TSCA : Yes

Documents