missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Phenetidine Hydrochloride 98.0+%, TCI America™

$77.92 - $557.66

Chemical Identifiers

CAS 637-56-9
Molecular Formula C8H12ClNO
Molecular Weight (g/mol) 173.64
MDL Number MFCD00035503
InChI Key JVWYCUBGJVOEIZ-UHFFFAOYSA-N
Synonym 4-Ethoxyaniline Hydrochloride
PubChem CID 69478
IUPAC Name 4-ethoxyaniline;hydrochloride
SMILES CCOC1=CC=C(C=C1)N.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
P011625G View Documents TCI America
P011625G
25g Each for $77.92
 
P0116500G View Documents TCI America
P0116500G
500g Each for $557.66
 
Specifications

Chemical Identifiers

637-56-9
173.64
JVWYCUBGJVOEIZ-UHFFFAOYSA-N
69478
CCOC1=CC=C(C=C1)N.Cl
C8H12ClNO
MFCD00035503
4-Ethoxyaniline Hydrochloride
4-ethoxyaniline;hydrochloride

Specifications

Red-White
637-56-9
MFCD00035503
JVWYCUBGJVOEIZ-UHFFFAOYSA-N
4-ethoxyaniline;hydrochloride
69478
Crystal-Powder at 20°C
p-Phenetidine Hydrochloride
25g
C8H12ClNO
4-Ethoxyaniline Hydrochloride
CCOC1=CC=C(C=C1)N.Cl
173.64
173.64
≥98.0% (HPLC,T)
Safety and Handling

Safety and Handling

RTECSNumber : SI7051000

TSCA : Yes