Please login to your online account to display your discounted pricing

p-Phenetidine 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  130200050

 View more versions of this product

Catalog No. AC130200050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Quantity 5g
Color Clear brown
Health Hazard 1 GHS Signal Word: Warning
EINECS Number 205-855-5
TSCA TSCA
RTECS Number SI6465500
Formula Weight 137.18
MDL Number MFCD00007865
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material p-Phenetidine
Infrared Spectrum Authentic
Name Note 98%
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C8H11NO
Refractive Index 1.5590 to 1.5610 (20°C, 589nm)
Linear Formula C2H5OC6H4NH2
CAS 156-43-4
Synonym 4-Aminoethoxybenzene
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
May cause an allergic skin reaction.
Suspected of causing genetic defects.
Harmful if inhaled.
Density 1.0600g/mL

Specifications

Chemical Name or Material p-Phenetidine
Name Note 98%
CAS 156-43-4
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H11NO
Linear Formula C2H5OC6H4NH2
Formula Weight 137.18
MDL Number MFCD00007865
Synonym 4-Aminoethoxybenzene
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
May cause an allergic skin reaction.
Suspected of causing genetic defects.
Harmful if inhaled.

SDS & Safety Documents