Please login to your online account to display your discounted pricing

p-Phenetidine 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material p-Phenetidine
Name Note 98%
CAS 156-43-4
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H11NO
Linear Formula C2H5OC6H4NH2
Formula Weight 137.18
MDL Number MFCD00007865
Synonym 4-Aminoethoxybenzene
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC130200050

  SDS
thermo fisher scientific
130200050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Quantity 5g
Color Clear brown
Health Hazard 1 GHS Signal Word: Warning
TSCA TSCA
RTECS Number SI6465500
EINECS Number 205-855-5
Formula Weight 137.18
MDL Number MFCD00007865
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
May cause an allergic skin reaction.
Suspected of causing genetic defects.
Harmful if inhaled.
Chemical Name or Material p-Phenetidine
Infrared Spectrum Authentic
Linear Formula C2H5OC6H4NH2
CAS 156-43-4
Synonym 4-Aminoethoxybenzene
Assay Percent Range 98%
Refractive Index 1.5590 to 1.5610 (20°C, 589nm)
Name Note 98%
Molecular Formula C8H11NO
Packaging Glass bottle
Density 1.0600g/mL
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
May cause an allergic skin reaction.
Suspected of causing genetic defects.
Harmful if inhaled.