Please login to your online account to display your discounted pricing

Alfa Aesar™ p-Phenetidine, 98%

Chemical Identifiers

Chemical Name or Material p-Phenetidine
CAS 156-43-4
Molecular Formula C8H11NO
Formula Weight 137.18
MDL Number MFCD00007865
UN Number UN2311
Beilstein 606666
Merck Index 14,7228
Synonym 4-Ethoxyaniline; 4-Aminophenetole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA104160E

 
alfa aesar™
A104160E
2500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1041636

 
alfa aesar™
A1041636
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 137.18
Chemical Name or Material p-Phenetidine
TSCA TSCA
Assay 98%
Molecular Formula C8H11NO
MDL Number MFCD00007865
CAS 156-43-4
UN Number UN2311
Beilstein 606666
Merck Index 14,7228
Synonym 4-Ethoxyaniline; 4-Aminophenetole
Odor Unpleasant
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H341-H302-H312-H332-H319-H317
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 254°C
Melting Point 2° to 4°C
Quantity 2500g
Flash Point 115°C (239°F)
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: PHENETIDINES
RTECS Number SI6465500

Applications
This chemical is mainly used as intermediate for several materials also used as medicine, dye, food preservatives, feed additives and rubber age-resistant intermediates.

Solubility
Not miscible or difficult to mix in water.

Notes
Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Store in dark.

Safety and Handling

Warning

GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H341-H302-H312-H332-H319-H317
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
May cause an allergic skin reaction.