Please login to your online account to display your discounted pricing

p-Nitrophenyl Phosphate (Disodium Salt, Hexahydrate), Fisher BioReagents


Quantity 5g
Packaging Amber Glass

Manufacturer: fisher scientific company  BP25345

 View more versions of this product

Catalog No. BP25345

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Melting Point 300°C
Physical Form Powder/Solid
Formula Weight 371
Quantity 5g
Molecular Formula C6H4NO6PNa2 · 6 H2O
Color Off-white
Chemical Name or Material p-Nitrophenyl Phosphate Disodium Salt, Hexahydrate
Water 27 to 30%
ChemAlert Storage Symbol Gray
Identification Pass Test
Packaging Amber Glass
Assay Percent Range ≥98.0%
CAS 4264-83-9
Recommended Storage Store below 0°C
Free p_Nitrophenol 0.01% max.

Specifications

Chemical Name or Material p-Nitrophenyl Phosphate Disodium Salt, Hexahydrate
Assay Percent Range ≥98.0%
Molecular Formula C6H4NO6PNa2 · 6 H2O
Formula Weight 371
Accessories
 

Store below 0°C

SDS & Safety Documents