missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Nitrophenyl Phosphate (Disodium Salt, Hexahydrate), Fisher BioReagents

$988.35 - $988.35

Chemical Identifiers

CAS 4264-83-9
Molecular Formula C6H4NNa2O6P
Molecular Weight (g/mol) 263.05
MDL Number MFCD00007319
InChI Key VIYFPAMJCJLZKD-UHFFFAOYSA-L
Synonym disodium 4-nitrophenyl phosphate, disodium 4-nitrophenylphosphate, disodium p-nitrophenyl phosphate, p-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, disodium salt, phosphoric acid, mono 4-nitrophenyl ester, sodium salt 1:2, pnpp, pnpp liquid substrate system, sodium 4-nitrophenyl phosphate
PubChem CID 77949
IUPAC Name disodium;(4-nitrophenyl) phosphate
SMILES [Na+].[Na+].[O-][N+](=O)C1=CC=C(OP([O-])([O-])=O)C=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BP253410
SDS View Documents Promotion Details
Fisher BioReagents
BP253410
10g Amber Glass
Each for $988.35
Case of 5 Each for $4,941.75
 
Description

Description

p-Nitrophenyl Phosphate is suitable for use as a substrate for alkaline and acid phosphatase.
Specifications

Chemical Identifiers

4264-83-9
263.05
VIYFPAMJCJLZKD-UHFFFAOYSA-L
77949
[Na+].[Na+].[O-][N+](=O)C1=CC=C(OP([O-])([O-])=O)C=C1
C6H4NNa2O6P
MFCD00007319
disodium 4-nitrophenyl phosphate, disodium 4-nitrophenylphosphate, disodium p-nitrophenyl phosphate, p-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, disodium salt, phosphoric acid, mono 4-nitrophenyl ester, sodium salt 1:2, pnpp, pnpp liquid substrate system, sodium 4-nitrophenyl phosphate
disodium;(4-nitrophenyl) phosphate

Specifications

4264-83-9
300°C
≥98.0%
MFCD00007319
10g
VIYFPAMJCJLZKD-UHFFFAOYSA-L
disodium;(4-nitrophenyl) phosphate
77949
Powder/Solid
p-Nitrophenyl Phosphate Disodium Salt, Hexahydrate
White
Pass Test
C6H4NNa2O6P
Amber Glass
disodium 4-nitrophenyl phosphate, disodium 4-nitrophenylphosphate, disodium p-nitrophenyl phosphate, p-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, disodium salt, phosphoric acid, mono 4-nitrophenyl ester, sodium salt 1:2, pnpp, pnpp liquid substrate system, sodium 4-nitrophenyl phosphate
[Na+].[Na+].[O-][N+](=O)C1=CC=C(OP([O-])([O-])=O)C=C1
263.05
371
27 to 30%
Safety and Handling

Safety and Handling

Recommended Storage : Store below 0°C

SDS
Product Certifications