missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing

p-Nitrophenyl Phosphate (Disodium Salt, Hexahydrate), Fisher BioReagents
Manufacturer: Fisher BioReagents BP253410
Description
p-Nitrophenyl Phosphate is suitable for use as a substrate for alkaline and acid phosphatase.Specifications
p-Nitrophenyl Phosphate Disodium Salt, Hexahydrate | |
White | |
Pass Test | |
C6H4NNa2O6P | |
Amber Glass | |
10g | |
[Na+].[Na+].[O-][N+](=O)C1=CC=C(OP([O-])([O-])=O)C=C1 | |
263.05 | |
371 | |
27 to 30% |
4264-83-9 | |
300°C | |
≥98.0% | |
MFCD00007319 | |
disodium 4-nitrophenyl phosphate, disodium 4-nitrophenylphosphate, disodium p-nitrophenyl phosphate, p-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, disodium salt, phosphoric acid, mono 4-nitrophenyl ester, sodium salt 1:2, pnpp, pnpp liquid substrate system, sodium 4-nitrophenyl phosphate | |
VIYFPAMJCJLZKD-UHFFFAOYSA-L | |
disodium;(4-nitrophenyl) phosphate | |
77949 | |
Powder/Solid |
Chemical Identifiers
4264-83-9 | |
263.05 | |
VIYFPAMJCJLZKD-UHFFFAOYSA-L | |
77949 | |
[Na+].[Na+].[O-][N+](=O)C1=CC=C(OP([O-])([O-])=O)C=C1 |
C6H4NNa2O6P | |
MFCD00007319 | |
disodium 4-nitrophenyl phosphate, disodium 4-nitrophenylphosphate, disodium p-nitrophenyl phosphate, p-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, disodium salt, phosphoric acid, mono 4-nitrophenyl ester, sodium salt 1:2, pnpp, pnpp liquid substrate system, sodium 4-nitrophenyl phosphate | |
disodium;(4-nitrophenyl) phosphate |
Safety and Handling

Recommended Storage : Store below 0°C