missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing

p-Nitrophenol (Yellow Crystals or Powder/Peptide Synthesis), Fisher BioReagents
Manufacturer: Fisher BioReagents FLBP6121
Description
Nitrophenol is commonly used in peptide synthesis procedures.Specifications
Nitrophenol | |
100-02-7 | |
Brown | |
279°C | |
≥99 % | |
MFCD00007331 | |
15, 6708 | |
1kg | |
OC1=CC=C(C=C1)[N+]([O-])=O | |
139.11 | |
CHEBI:16836 | |
Solid | |
Peptide Synthesis |
p-Nitrophenol | |
4.4 | |
112°C | |
Pass Test | |
C6H5NO3 | |
Glass Bottle | |
4-hydroxynitrobenzene, mononitrophenol, niphen, p-hydroxynitrobenzene, p-nitrophenol, paranitrofenol, paranitrofenolo, paranitrophenol, phenol, 4-nitro, phenol, p-nitro | |
BTJIUGUIPKRLHP-UHFFFAOYSA-N | |
4-nitrophenol | |
980 | |
139.11 | |
≥99% |
Chemical Identifiers
100-02-7 | |
139.11 | |
BTJIUGUIPKRLHP-UHFFFAOYSA-N | |
980 | |
4-nitrophenol |
C6H5NO3 | |
MFCD00007331 | |
4-hydroxynitrobenzene, mononitrophenol, niphen, p-hydroxynitrobenzene, p-nitrophenol, paranitrofenol, paranitrofenolo, paranitrophenol, phenol, 4-nitro, phenol, p-nitro | |
CHEBI:16836 | |
OC1=CC=C(C=C1)[N+]([O-])=O |
Safety and Handling

Recommended Storage : RT