missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Nitroacetophenone, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 128341000

 View more versions of this product

Catalog No. AC128341000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

p-Nitroacetophenone
100-19-6
Yellow
Authentic
Glass bottle
O2NC6H4COCH3
100 g
p-nitroacetophenone, 4'-nitroacetophenone, 4-nitroacetophenone, 1-4-nitrophenyl ethanone, p-acetylnitrobenzene, paranitroacetophenone, p-nitrophenyl methyl ketone, 1-4-nitrophenyl ethan-1-one, ethanone, 1-4-nitrophenyl, pnap
CC(=O)C1=CC=C(C=C1)[N+]([O-])=O
165.15
CHEBI:28735
97%
97%
76°C to 80°C
202°C
97%
C8H7NO3
MFCD00007355
07, 288
YQYGPGKTNQNXMH-UHFFFAOYSA-N
1-(4-nitrophenyl)ethanone
7487
165.15
Crystalline Powder

Chemical Identifiers

100-19-6
165.15
YQYGPGKTNQNXMH-UHFFFAOYSA-N
7487
1-(4-nitrophenyl)ethanone
C8H7NO3
MFCD00007355
p-nitroacetophenone, 4'-nitroacetophenone, 4-nitroacetophenone, 1-4-nitrophenyl ethanone, p-acetylnitrobenzene, paranitroacetophenone, p-nitrophenyl methyl ketone, 1-4-nitrophenyl ethan-1-one, ethanone, 1-4-nitrophenyl, pnap
CHEBI:28735
CC(=O)C1=CC=C(C=C1)[N+]([O-])=O
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-827-4

RTECSNumber : AM9627000

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only