missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Naphtholbenzein OR, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 145-50-6
Molecular Formula 4-HOC10H6C(C6H5):C10H6-4-:O
Formula Weight 374.44
Chemical Name or Material p-Naphtholbenzein
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MKE12110 Avantor Performance
E12154
Glass 10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

145-50-6
4-HOC10H6C(C6H5):C10H6-4-:O
374.44
p-Naphtholbenzein

Specifications

374.44
4-HOC10H6C(C6H5):C10H6-4-:O
Glass
10g
145-50-6
p-Naphtholbenzein