Learn More
p-Hydroxycinnamic acid, 98%, predominantly trans, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 121090250
Specifications
p-Hydroxycinnamic acid | |
Plastic bottle | |
214°C | |
98% | |
97.5% min. (HPLC) | |
HOC6H4CH=CHCO2H | |
10, 297 | |
3-4-hydroxyphenyl acrylic acid, 4-coumaric acid, 4-hydroxycinnamic acid, hydroxycinnamic acid, p-coumaric acid, p-cumaric acid, p-hydroxycinnamic acid, para-coumaric acid, trans-4-hydroxycinnamic acid, trans-p-coumaric acid | |
NGSWKAQJJWESNS-ZZXKWVIFSA-N | |
(E)-3-(4-hydroxyphenyl)prop-2-enoic acid | |
637542 | |
164.16 | |
98% |
Authentic | |
Beige to White or Green | |
25g | |
501-98-4 | |
C9H8O3 | |
MFCD00004399 | |
14, 2560 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol and ether,practically insoluble in benzene and ligroine | |
C1=CC(=CC=C1C=CC(=O)O)O | |
164.16 | |
CHEBI:32374 | |
Powder |
Chemical Identifiers
501-98-4 | |
164.16 | |
NGSWKAQJJWESNS-ZZXKWVIFSA-N | |
637542 | |
(E)-3-(4-hydroxyphenyl)prop-2-enoic acid |
C9H8O3 | |
MFCD00004399 | |
3-4-hydroxyphenyl acrylic acid, 4-coumaric acid, 4-hydroxycinnamic acid, hydroxycinnamic acid, p-coumaric acid, p-cumaric acid, p-hydroxycinnamic acid, para-coumaric acid, trans-4-hydroxycinnamic acid, trans-p-coumaric acid | |
CHEBI:32374 | |
C1=CC(=CC=C1C=CC(=O)O)O |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning