missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
p-Fluorotoluene, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 119672500
Specifications
p-Fluorotoluene | |
10°C | |
Glass bottle | |
1 | |
116°C | |
-56°C | |
97% | |
97% | |
CH3C6H4F | |
05, 290 | |
1-methyl-4-fluorobenzene, 4-fluorotoluene, benzene, 1-fluoro-4-methyl, p-fluoromethyl benzene, p-fluoromethylbenzene, p-fluorotoluene, p-tolyl fluoride, para-fluorotoluene, toluene, p-fluoro, unii-nud6wz1b8f | |
WRWPPGUCZBJXKX-UHFFFAOYSA-N | |
1-fluoro-4-methylbenzene | |
9603 | |
Liquid |
1.0000g/mL | |
Authentic | |
1.4674 to 1.4694 | |
0.05% max. (K.F.) | |
Colorless to Yellow | |
250g | |
352-32-9 | |
C7H7F | |
MFCD00000358 | |
15, 4212 | |
Solubility in water: insoluble.,soluble in alcohols,ether | |
CC1=CC=C(C=C1)F | |
110.13 | |
110.13 | |
97% |
Chemical Identifiers
352-32-9 | |
110.13 | |
WRWPPGUCZBJXKX-UHFFFAOYSA-N | |
9603 | |
CC1=CC=C(C=C1)F |
C7H7F | |
MFCD00000358 | |
1-methyl-4-fluorobenzene, 4-fluorotoluene, benzene, 1-fluoro-4-methyl, p-fluoromethyl benzene, p-fluoromethylbenzene, p-fluorotoluene, p-tolyl fluoride, para-fluorotoluene, toluene, p-fluoro, unii-nud6wz1b8f | |
1-fluoro-4-methylbenzene |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
If eye irritation persists: Get medical advice/attention.
GHS Signal Word: Danger
EINECSNumber : 206-520-6
RTECSNumber : XT2580000
TSCA : TSCA