missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ p-Fluoranil, 97%

$63.23 - $223.61

Chemical Identifiers

CAS 527-21-9
Molecular Formula C6F4O2
Molecular Weight (g/mol) 180.058
MDL Number MFCD00001592
InChI Key JKLYZOGJWVAIQS-UHFFFAOYSA-N
Synonym tetrafluoro-1,4-benzoquinone, fluoranil, p-fluoranil, tetrafluoroquinone, tetrafluoro-p-benzoquinone, fluoroanil, tetrafluorocyclohexa-2,5-diene-1,4-dione, 2,5-cyclohexadiene-1,4-dione, 2,3,5,6-tetrafluoro, 2,3,5,6-tetrafluoro-1,4-benzoquinone, 2,3,5,6-tetrafluoro 1,4 benzoquinone
PubChem CID 68239
IUPAC Name 2,3,5,6-tetrafluorocyclohexa-2,5-diene-1,4-dione
SMILES C1(=C(C(=O)C(=C(C1=O)F)F)F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0128703 View Documents Alfa Aesar™
L0128703
1g Each for $63.23
 
AAL0128706 View Documents Alfa Aesar™
L0128706
5g Each for $223.61
 
Specifications

Chemical Identifiers

527-21-9
180.058
JKLYZOGJWVAIQS-UHFFFAOYSA-N
68239
C1(=C(C(=O)C(=C(C1=O)F)F)F)F
C6F4O2
MFCD00001592
tetrafluoro-1,4-benzoquinone, fluoranil, p-fluoranil, tetrafluoroquinone, tetrafluoro-p-benzoquinone, fluoroanil, tetrafluorocyclohexa-2,5-diene-1,4-dione, 2,5-cyclohexadiene-1,4-dione, 2,3,5,6-tetrafluoro, 2,3,5,6-tetrafluoro-1,4-benzoquinone, 2,3,5,6-tetrafluoro 1,4 benzoquinone
2,3,5,6-tetrafluorocyclohexa-2,5-diene-1,4-dione

Specifications

~184°C (sublimation)
527-21-9
MFCD00001592
tetrafluoro-1,4-benzoquinone, fluoranil, p-fluoranil, tetrafluoroquinone, tetrafluoro-p-benzoquinone, fluoroanil, tetrafluorocyclohexa-2,5-diene-1,4-dione, 2,5-cyclohexadiene-1,4-dione, 2,3,5,6-tetrafluoro, 2,3,5,6-tetrafluoro-1,4-benzoquinone, 2,3,5,6-tetrafluoro 1,4 benzoquinone
C1(=C(C(=O)C(=C(C1=O)F)F)F)F
180.058
180.06
p-Fluoranil
1g
C6F4O2
1875039
JKLYZOGJWVAIQS-UHFFFAOYSA-N
2,3,5,6-tetrafluorocyclohexa-2,5-diene-1,4-dione
68239
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning